CH4244052455 ChemBridge Corporation

5-amino-2-fluorobenzamide

  • MDL number MFCD03094256
  • Cas Number : 518057-72-2
  • Molecular Weight : 154.1417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036164

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036164
Availability details for
Quantity
  • Availability
  • Add to Cart