CH4244052508 ChemBridge Corporation

6-chloro-N~4~-methyl-2,4-pyrimidinediamine

  • MDL number MFCD02091108
  • Cas Number : 1005-37-4
  • Molecular Weight : 158.5889

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041389

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4041389
Availability details for
Quantity
  • Availability
  • Add to Cart