CH4244052509 ChemBridge Corporation

1-phenyl-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazine

  • MDL number MFCD00297430
  • Cas Number : 112758-89-1
  • Molecular Weight : 198.2637

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100076

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4100076
Availability details for
Quantity
  • Availability
  • Add to Cart