CH4244052514 ChemBridge Corporation

2-(4-chloro-3,5-dimethyl-1H-pyrazol-1-yl)ethanamine

  • MDL number MFCD04967723
  • Cas Number : 925634-46-4
  • Molecular Weight : 173.6433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101874

Safety Information

MSDS https://www.hit2lead.com/sds/4101874_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart