CH4244052527 ChemBridge Corporation

6-chloro-N~4~,N~4~-dimethyl-2,4-pyrimidinediamine

  • MDL number MFCD13188630
  • Cas Number : 1007-11-0
  • Molecular Weight : 172.6155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9071107

Safety Information

MSDS https://www.hit2lead.com/building-blocks/9071107
Availability details for
Quantity
  • Availability
  • Add to Cart