CH4250699451 ChemBridge Corporation

1-benzyl-3-bromo-N,N-dimethyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD14702897
  • Cas Number : 1215296-00-6
  • Molecular Weight : 281.1517

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040160

Safety Information

MSDS https://www.hit2lead.com/sds/4040160_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart