CH4250699453 ChemBridge Corporation

(3-bromo-1H-1,2,4-triazol-5-yl)methanol

  • MDL number MFCD14702904
  • Cas Number : 23164-67-2
  • Molecular Weight : 177.9874

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040469

Safety Information

MSDS https://www.hit2lead.com/sds/4040469_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart