CH4250699459 ChemBridge Corporation

5-amino-2-hydroxy-N,N-dimethylbenzamide

  • MDL number MFCD14702915
  • Cas Number : 862853-55-2
  • Molecular Weight : 180.2038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041000

Safety Information

MSDS https://www.hit2lead.com/sds/4041000_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart