CH4250699524 ChemBridge Corporation

2-oxo-1,2-dihydro-3-quinolinecarboxylic acid

  • MDL number MFCD01550054
  • Cas Number : 2003-79-4
  • Molecular Weight : 189.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009893

Safety Information

MSDS https://www.hit2lead.com/sds/4009893_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart