CH4250699526 ChemBridge Corporation

4-methyl-3-oxo-2,3-dihydro-1H-pyrazolo[3,4-b]pyridine-6-carboxylic acid

  • MDL number MFCD11982976
  • Cas Number : 862273-50-5
  • Molecular Weight : 193.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4035782

Safety Information

msds https://www.hit2lead.com/building-blocks/4035782
Availability details for
Quantity
  • Availability
  • Add to Cart