CH4250699536 ChemBridge Corporation

(4-amino-2-methylphenyl)dimethylamine

  • MDL number MFCD09965938
  • Cas Number : 27746-11-8
  • Molecular Weight : 150.2209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036059

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036059
Availability details for
Quantity
  • Availability
  • Add to Cart