CH4250699554 ChemBridge Corporation

2-bromo-4,6-dimethylpyridine

  • MDL number MFCD00082591
  • Cas Number : 4926-26-5
  • Molecular Weight : 186.0491

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036526

Safety Information

MSDS https://www.hit2lead.com/sds/4036526_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart