CH4250699567 ChemBridge Corporation

1H-pyrrolo[2,3-c]pyridine-5-carbaldehyde

  • MDL number MFCD10697530
  • Cas Number : 130473-26-6
  • Molecular Weight : 146.146

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036705

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036705
Availability details for
Quantity
  • Availability
  • Add to Cart