CH4250699583 ChemBridge Corporation

4-oxatricyclo[4.3.1.1~3,8~]undecan-5-one

  • MDL number MFCD00184584
  • Cas Number : 21898-84-0
  • Molecular Weight : 166.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036791

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036791
Availability details for
Quantity
  • Availability
  • Add to Cart