CH4250699616 ChemBridge Corporation

1-methyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylic acid

  • MDL number MFCD09901219
  • Cas Number : 32286-99-0
  • Molecular Weight : 180.2038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017570

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4017570
Availability details for
Quantity
  • Availability
  • Add to Cart