CH4250699630 ChemBridge Corporation

1-ethyl-1H-pyrazole-3-carbonitrile

  • MDL number MFCD04969701
  • Cas Number : 1006471-40-4
  • Molecular Weight : 121.1399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040120

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4040120
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart