CH4250699637 ChemBridge Corporation

4-ethoxy-1-naphthalenesulfonyl chloride

  • MDL number MFCD09044337
  • Cas Number : 91222-55-8
  • Molecular Weight : 270.7319

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040239

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4040239_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart