CH4250699639 ChemBridge Corporation

3-hydrazino-1-methyl-2(1H)-quinoxalinone

  • MDL number MFCD14702900
  • Cas Number : 31595-64-9
  • Molecular Weight : 190.2019

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040244

Safety Information

MSDS https://www.hit2lead.com/sds/4040244_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart