CH4250699649 ChemBridge Corporation

3,5-dichloro-1H-1,2,4-triazole

  • MDL number MFCD01421959
  • Cas Number : 10327-87-4
  • Molecular Weight : 137.9554

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040447

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040447
Availability details for
Quantity
  • Availability
  • Add to Cart