CH4250699697 ChemBridge Corporation

1-(2-pyridinyl)-2-propanol

  • MDL number MFCD01691728
  • Cas Number : 5307-19-7
  • Molecular Weight : 137.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041318

Safety Information

MSDS https://www.hit2lead.com/sds/4041318_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart