CH4250699725 ChemBridge Corporation

2-ethyl-1H-benzimidazol-6-amine

  • MDL number MFCD00462860
  • Cas Number : 46055-62-3
  • Molecular Weight : 161.2037

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5176078

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5176078
Availability details for
Quantity
  • Availability
  • Add to Cart