CH4301081765 ChemBridge Corporation

5-cyclopropyl-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06823674
  • Cas Number : 1287752-79-7
  • Molecular Weight : 166.1772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039906

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4039906
Availability details for
Quantity
  • Availability
  • Add to Cart