CH4301081766 ChemBridge Corporation

2-(4-chloro-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD04969792
  • Cas Number : 51363-82-7
  • Molecular Weight : 174.585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040008

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040008
Availability details for
Quantity
  • Availability
  • Add to Cart