CH4301081768 ChemBridge Corporation

3-(6-methyl-2-oxo-1(2H)-pyridinyl)propanoic acid

  • MDL number MFCD11911382
  • Cas Number : 859296-04-1
  • Molecular Weight : 181.1885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040106

Safety Information

MSDS https://www.hit2lead.com/sds/4040106_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart