CH4301081778 ChemBridge Corporation

2-cyclohexylpropanoic acid

  • MDL number MFCD00268172
  • Cas Number : 6051-13-4
  • Molecular Weight : 156.2221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040626

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040626
Availability details for
Quantity
  • Availability
  • Add to Cart