CH4301081780 ChemBridge Corporation

4-chloro-1,3-dimethyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02090838
  • Cas Number : 98198-65-3
  • Molecular Weight : 174.585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040780

Safety Information

MSDS https://www.hit2lead.com/sds/4040780_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart