CH4309656410 ChemBridge Corporation

tert-butyl rac-(1S,6R)-3,9-diazabicyclo[4.2.1]nonane-3-carboxylate

  • MDL number MFCD12755902
  • Cas Number : 1932112-98-5
  • Molecular Weight : 226.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031609

Safety Information

MSDS https://www.hit2lead.com/sds/4031609_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart