CH4309656413 ChemBridge Corporation

tert-butyl 3,9-diazaspiro[5.6]dodecane-3-carboxylate

  • MDL number MFCD18381890
  • Cas Number : 1256643-27-2
  • Molecular Weight : 268.395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036476

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4036476
Availability details for
Quantity
  • Availability
  • Add to Cart