CH4309656450 ChemBridge Corporation

6-chloro-2,3,4-trimethylpyridine

  • MDL number MFCD17078867
  • Cas Number : 408314-12-5
  • Molecular Weight : 155.6247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040134

Safety Information

MSDS https://www.hit2lead.com/sds/4040134_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart