CH4309656461 ChemBridge Corporation

5-bromo-2-ethyl-2,4-dihydro-3H-1,2,4-triazol-3-one

  • MDL number MFCD17078876
  • Cas Number : 1243250-10-3
  • Molecular Weight : 192.0139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040523

Safety Information

MSDS https://www.hit2lead.com/sds/4040523_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart