CH4309656475 ChemBridge Corporation

3,5-dichloro-1-ethyl-1H-1,2,4-triazole

  • MDL number MFCD18381920
  • Cas Number : 1256643-69-2
  • Molecular Weight : 166.0086

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4041202

Safety Information

MSDS https://www.hit2lead.com/sds/4041202_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart