CH4309656488 ChemBridge Corporation

4-iodo-3-methyl-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD18064539
  • Cas Number : 855343-07-6
  • Molecular Weight : 275.9983

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042704

Safety Information

MSDS https://www.hit2lead.com/sds/4042704_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart