CH4309656495 ChemBridge Corporation

4-amino-7-methylpyrazolo[5,1-c][1,2,4]triazine-3-carbonitrile

  • MDL number MFCD18381933
  • Cas Number : 63475-17-2
  • Molecular Weight : 174.1627

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4042928

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4042928_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart