CH4309656506 ChemBridge Corporation

1-(1,4-dimethyl-2-piperazinyl)methanamine

  • MDL number MFCD13151976
  • Cas Number : 131922-07-1
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015474

Safety Information

MSDS https://www.hit2lead.com/sds/4015474_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart