CH4309656533 ChemBridge Corporation

propoxyacetic acid

  • MDL number MFCD09744041
  • Cas Number : 54497-00-6
  • Molecular Weight : 118.1311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4020861

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4020861
Availability details for
Quantity
  • Availability
  • Add to Cart