CH4309656551 ChemBridge Corporation

1,6-dimethyl-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid

  • MDL number MFCD08361851
  • Cas Number : 889939-62-2
  • Molecular Weight : 167.162

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026285

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4026285
Availability details for
Quantity
  • Availability
  • Add to Cart