CH4309656560 ChemBridge Corporation

4-(5-methyl-1,2,4-oxadiazol-3-yl)phenol

  • MDL number MFCD06655819
  • Cas Number : 49787-02-2
  • Molecular Weight : 176.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026890

Safety Information

MSDS https://www.hit2lead.com/sds/4026890_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart