CH4309656615 ChemBridge Corporation

5-(methoxymethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01203002
  • Cas Number : 15884-86-3
  • Molecular Weight : 145.1829

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035319

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035319
Availability details for
Quantity
  • Availability
  • Add to Cart