CH4309656622 ChemBridge Corporation

2-cyclopropyl-6-oxo-1,6-dihydro-5-pyrimidinecarboxylic acid

  • MDL number MFCD18178868
  • Cas Number : 1219561-35-9
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035618

Safety Information

MSDS https://www.hit2lead.com/sds/4035618_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart