CH4309656630 ChemBridge Corporation

3-chloro-5-isoxazolecarboxylic acid

  • MDL number MFCD01716783
  • Cas Number : 20724-56-5
  • Molecular Weight : 147.5166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036344

Safety Information

MSDS https://www.hit2lead.com/sds/4036344_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart