CH4309656641 ChemBridge Corporation

2-ethylisonicotinic acid

  • MDL number MFCD09910286
  • Cas Number : 3376-96-3
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036495

Safety Information

MSDS https://www.hit2lead.com/sds/4036495_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart