CH4309656673 ChemBridge Corporation

isoxazole-3-carbaldehyde

  • MDL number MFCD06739019
  • Cas Number : 89180-61-0
  • Molecular Weight : 97.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002199

Safety Information

MSDS https://www.hit2lead.com/sds/4002199_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart