CH4309656677 ChemBridge Corporation

2-bromo-6-isopropylpyridine

  • MDL number MFCD11223220
  • Cas Number : 1037223-35-0
  • Molecular Weight : 200.0757

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4037569

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4037569
Availability details for
Quantity
  • Availability
  • Add to Cart