CH4309656685 ChemBridge Corporation

5,6-dimethyl-4-pyrimidinamine

  • MDL number MFCD09909808
  • Cas Number : 54568-12-6
  • Molecular Weight : 123.1558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4037920

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4037920
Availability details for
Quantity
  • Availability
  • Add to Cart