CH4309656705 ChemBridge Corporation

6-ethyl-4-pyrimidinol

  • MDL number MFCD09909797
  • Cas Number : 124703-78-2
  • Molecular Weight : 124.1405

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038765

Safety Information

MSDS https://www.hit2lead.com/sds/4038765_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart