CH4309656713 ChemBridge Corporation

1,5-dimethyl-1H-indole-2-carboxylic acid

  • MDL number MFCD03848206
  • Cas Number : 216210-59-2
  • Molecular Weight : 189.2105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002670

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4002670
Availability details for
Quantity
  • Availability
  • Add to Cart