CH4309656757 ChemBridge Corporation

[3-(chlorosulfonyl)-4-methoxyphenyl]acetic acid

  • MDL number MFCD09050452
  • Cas Number : 104967-35-3
  • Molecular Weight : 264.6828

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039911

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/building-blocks/4039911
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart