CH4309656762 ChemBridge Corporation

2-isobutyl-1H-imidazole

  • MDL number MFCD15146457
  • Cas Number : 61491-92-7
  • Molecular Weight : 124.1836

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039941

Safety Information

MSDS https://www.hit2lead.com/sds/4039941_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart