CH4309656768 ChemBridge Corporation

3-bromo-1H-1,2,4-triazole-5-carboxamide

  • MDL number MFCD16556127
  • Cas Number : 1207755-03-0
  • Molecular Weight : 190.9861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039970

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4039970
Availability details for
Quantity
  • Availability
  • Add to Cart