CH4309656770 ChemBridge Corporation

ethyl 4-amino-1,2,5-oxadiazole-3-carboxylate

  • MDL number MFCD00465050
  • Cas Number : 17376-63-5
  • Molecular Weight : 157.1274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039986

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4039986
Availability details for
Quantity
  • Availability
  • Add to Cart